The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002910.5 | NP_002901.2 | 6 - 7 | 850 | 57 |
XM_017029698.1 | XP_016885187.1 | 6 - 7 | 745 | 57 | |
GenBank mRNA | D10232.1 | - | 6 - 7 | 680 | 57 |