The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178564.3 | NP_848659.2 | 2 - 3 | 395 | 93 |
XM_017013378.1 | XP_016868867.1 | 2 - 3 | 395 | 93 | |
XM_017013379.1 | XP_016868868.1 | 2 - 3 | 395 | 93 | |
XM_017013380.1 | XP_016868869.1 | 2 - 3 | 395 | 93 | |
XM_017013381.1 | XP_016868870.1 | 2 - 3 | 395 | 93 | |
XM_017013382.1 | XP_016868871.1 | 2 - 3 | 395 | 93 | |
XM_017013383.1 | XP_016868872.1 | 2 - 3 | 395 | 93 | |
GenBank mRNA | AK293943.1 | - | 2 - 3 | 395 | 93 |
AL137662.1 | - | 1 - 2 | 138 | 93 | |
BC113873.1 | - | 1 - 2 | 110 | 93 | |
BC114481.1 | - | 1 - 2 | 110 | 93 |