Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NRBP2
Entrez Gene ID
340371
Gene Name
nuclear receptor binding protein 2
Gene Aliases
TRG16, pp9320
Chromosome Location
Chr.8: 143829776 - 143840974 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178564.3 NP_848659.2 3 - 4 496 68
XM_017013378.1 XP_016868867.1 3 - 4 496 68
XM_017013379.1 XP_016868868.1 3 - 4 496 68
XM_017013380.1 XP_016868869.1 3 - 4 496 68
XM_017013381.1 XP_016868870.1 3 - 4 496 68
XM_017013382.1 XP_016868871.1 3 - 4 496 68
XM_017013383.1 XP_016868872.1 3 - 4 496 68
GenBank mRNA AK293943.1 - 3 - 4 496 68
AL137662.1 - 2 - 3 239 68
BC071605.1 - 3 - 4 473 68
BC114481.1 - 2 - 3 211 68

View Full Product Details