Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYCBP2
Entrez Gene ID
23077
Gene Name
MYC binding protein 2, E3 ubiquitin protein ligase
Gene Aliases
Myc-bp2, PAM, Phr
Chromosome Location
Chr.13: 77044657 - 77327066 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015057.4 NP_055872.4 19 - 20 3124 89
XM_005266299.2 XP_005266356.1 19 - 20 3148 89
XM_006719779.2 XP_006719842.1 19 - 20 3148 89
XM_006719780.2 XP_006719843.1 19 - 20 3148 89
XM_006719781.2 XP_006719844.1 19 - 20 3148 89
XM_006719782.2 XP_006719845.1 19 - 20 3148 89
XM_006719783.2 XP_006719846.1 19 - 20 3148 89
XM_006719784.2 XP_006719847.1 18 - 19 3040 89
XM_006719785.2 XP_006719848.1 19 - 20 3148 89
XM_006719786.2 XP_006719849.1 19 - 20 3148 89
XM_006719787.2 XP_006719850.1 19 - 20 3148 89
XM_006719788.2 XP_006719851.1 19 - 20 3148 89
XM_006719789.2 XP_006719852.1 19 - 20 3148 89
XM_011535004.2 XP_011533306.1 18 - 19 2687 89
XM_011535005.2 XP_011533307.1 2 - 3 228 89
XM_017020454.1 XP_016875943.1 19 - 20 3148 89
XM_017020455.1 XP_016875944.1 19 - 20 3148 89
XM_017020456.1 XP_016875945.1 19 - 20 3148 89
XM_017020457.1 XP_016875946.1 19 - 20 3148 89
XM_017020458.1 XP_016875947.1 18 - 19 3040 89
XM_017020459.1 XP_016875948.1 18 - 19 3040 89
XM_017020460.1 XP_016875949.1 19 - 20 3148 89
XM_017020461.1 XP_016875950.1 19 - 20 3148 89
XM_017020462.1 XP_016875951.1 19 - 20 3148 89
XM_017020463.1 XP_016875952.1 18 - 19 3040 89
XM_017020464.1 XP_016875953.1 19 - 20 3148 89
XM_017020465.1 XP_016875954.1 18 - 19 3040 89
XM_017020466.1 XP_016875955.1 19 - 20 3148 89
XM_017020467.1 XP_016875956.1 19 - 20 3148 89
XM_017020468.1 XP_016875957.1 18 - 19 2635 89
GenBank mRNA AF075587.1 - 20 - 21 2889 89
BX473679.1 - 2 - 3 190 89

View Full Product Details