Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MYCBP2
- Entrez Gene ID
-
23077
- Gene Name
- MYC binding protein 2, E3 ubiquitin protein ligase
- Gene Aliases
-
Myc-bp2, PAM, Phr
- Chromosome Location
-
Chr.13: 77044657 - 77327066 on Build GRCh38
View Full Product Details