Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYCBP2
Entrez Gene ID
23077
Gene Name
MYC binding protein 2, E3 ubiquitin protein ligase
Gene Aliases
Myc-bp2, PAM, Phr
Chromosome Location
Chr.13: 77044657 - 77327066 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015057.4 NP_055872.4 48 - 49 7453 145
XM_005266299.2 XP_005266356.1 48 - 49 7477 145
XM_006719779.2 XP_006719842.1 48 - 49 7477 145
XM_006719780.2 XP_006719843.1 48 - 49 7477 145
XM_006719781.2 XP_006719844.1 48 - 49 7477 145
XM_006719782.2 XP_006719845.1 48 - 49 7477 145
XM_006719783.2 XP_006719846.1 48 - 49 7477 145
XM_006719784.2 XP_006719847.1 47 - 48 7369 145
XM_006719785.2 XP_006719848.1 48 - 49 7477 145
XM_006719786.2 XP_006719849.1 48 - 49 7477 145
XM_006719787.2 XP_006719850.1 48 - 49 7477 145
XM_006719788.2 XP_006719851.1 48 - 49 7477 145
XM_006719789.2 XP_006719852.1 48 - 49 7477 145
XM_011535004.2 XP_011533306.1 47 - 48 7016 145
XM_011535005.2 XP_011533307.1 31 - 32 4557 145
XM_017020454.1 XP_016875943.1 48 - 49 7477 145
XM_017020455.1 XP_016875944.1 48 - 49 7477 145
XM_017020456.1 XP_016875945.1 48 - 49 7477 145
XM_017020457.1 XP_016875946.1 48 - 49 7477 145
XM_017020458.1 XP_016875947.1 47 - 48 7369 145
XM_017020459.1 XP_016875948.1 47 - 48 7369 145
XM_017020460.1 XP_016875949.1 48 - 49 7477 145
XM_017020461.1 XP_016875950.1 48 - 49 7477 145
XM_017020462.1 XP_016875951.1 48 - 49 7477 145
XM_017020463.1 XP_016875952.1 47 - 48 7369 145
XM_017020464.1 XP_016875953.1 48 - 49 7477 145
XM_017020465.1 XP_016875954.1 47 - 48 7369 145
XM_017020466.1 XP_016875955.1 48 - 49 7477 145
XM_017020467.1 XP_016875956.1 48 - 49 7477 145
XM_017020468.1 XP_016875957.1 47 - 48 6964 145
GenBank mRNA AF075587.1 - 49 - 50 7218 145

View Full Product Details