Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYCBP2
Entrez Gene ID
23077
Gene Name
MYC binding protein 2, E3 ubiquitin protein ligase
Gene Aliases
Myc-bp2, PAM, Phr
Chromosome Location
Chr.13: 77044657 - 77327066 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015057.4 NP_055872.4 60 - 61 10796 110
XM_005266299.2 XP_005266356.1 62 - 63 11051 110
XM_006719779.2 XP_006719842.1 63 - 64 11177 110
XM_006719780.2 XP_006719843.1 63 - 64 11177 110
XM_006719781.2 XP_006719844.1 63 - 64 11153 110
XM_006719782.2 XP_006719845.1 62 - 63 11102 110
XM_006719783.2 XP_006719846.1 62 - 63 11075 110
XM_006719784.2 XP_006719847.1 62 - 63 11069 110
XM_006719785.2 XP_006719848.1 62 - 63 10997 110
XM_006719786.2 XP_006719849.1 61 - 62 10895 110
XM_006719787.2 XP_006719850.1 61 - 62 10877 110
XM_006719788.2 XP_006719851.1 61 - 62 10853 110
XM_006719789.2 XP_006719852.1 62 - 63 11051 110
XM_011535004.2 XP_011533306.1 62 - 63 10716 110
XM_011535005.2 XP_011533307.1 46 - 47 8257 110
XM_017020454.1 XP_016875943.1 63 - 64 11120 110
XM_017020455.1 XP_016875944.1 62 - 63 11045 110
XM_017020456.1 XP_016875945.1 62 - 63 10994 110
XM_017020457.1 XP_016875946.1 61 - 62 10976 110
XM_017020458.1 XP_016875947.1 61 - 62 10943 110
XM_017020459.1 XP_016875948.1 60 - 61 10787 110
XM_017020460.1 XP_016875949.1 60 - 61 10775 110
XM_017020461.1 XP_016875950.1 60 - 61 10763 110
XM_017020462.1 XP_016875951.1 60 - 61 10751 110
XM_017020463.1 XP_016875952.1 59 - 60 10712 110
XM_017020464.1 XP_016875953.1 60 - 61 10697 110
XM_017020465.1 XP_016875954.1 59 - 60 10643 110
XM_017020466.1 XP_016875955.1 59 - 60 10595 110
XM_017020467.1 XP_016875956.1 59 - 60 10595 110
XM_017020468.1 XP_016875957.1 62 - 63 10664 110
GenBank mRNA AB020723.1 - 1 - 2 126 110
AF075587.1 - 61 - 62 10564 110
BX647202.1 - 3 - 4 468 110

View Full Product Details