Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYCBP2
Entrez Gene ID
23077
Gene Name
MYC binding protein 2, E3 ubiquitin protein ligase
Gene Aliases
Myc-bp2, PAM, Phr
Chromosome Location
Chr.13: 77044657 - 77327066 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015057.4 NP_055872.4 73 - 74 12934 88
XM_005266299.2 XP_005266356.1 75 - 76 13189 88
XM_006719779.2 XP_006719842.1 76 - 77 13315 88
XM_006719780.2 XP_006719843.1 76 - 77 13306 88
XM_006719781.2 XP_006719844.1 76 - 77 13291 88
XM_006719782.2 XP_006719845.1 75 - 76 13240 88
XM_006719783.2 XP_006719846.1 75 - 76 13213 88
XM_006719784.2 XP_006719847.1 75 - 76 13207 88
XM_006719785.2 XP_006719848.1 75 - 76 13135 88
XM_006719786.2 XP_006719849.1 74 - 75 13033 88
XM_006719787.2 XP_006719850.1 74 - 75 13015 88
XM_006719788.2 XP_006719851.1 74 - 75 12991 88
XM_006719789.2 XP_006719852.1 75 - 76 13180 88
XM_011535004.2 XP_011533306.1 75 - 76 12854 88
XM_011535005.2 XP_011533307.1 59 - 60 10395 88
XM_017020454.1 XP_016875943.1 76 - 77 13258 88
XM_017020455.1 XP_016875944.1 75 - 76 13183 88
XM_017020456.1 XP_016875945.1 75 - 76 13132 88
XM_017020457.1 XP_016875946.1 74 - 75 13114 88
XM_017020458.1 XP_016875947.1 74 - 75 13081 88
XM_017020459.1 XP_016875948.1 73 - 74 12925 88
XM_017020460.1 XP_016875949.1 73 - 74 12913 88
XM_017020461.1 XP_016875950.1 73 - 74 12901 88
XM_017020462.1 XP_016875951.1 73 - 74 12889 88
XM_017020463.1 XP_016875952.1 72 - 73 12850 88
XM_017020464.1 XP_016875953.1 73 - 74 12835 88
XM_017020465.1 XP_016875954.1 72 - 73 12781 88
XM_017020466.1 XP_016875955.1 72 - 73 12733 88
XM_017020467.1 XP_016875956.1 72 - 73 12724 88
XM_017020468.1 XP_016875957.1 75 - 76 12802 88
GenBank mRNA AB020723.1 - 14 - 15 2264 88
AF075587.1 - 74 - 75 12702 88
AK025250.1 - 12 - 13 1814 88
AK025299.1 - 7 - 8 1038 88
BC037971.2 - 7 - 8 1201 88
BX647202.1 - 16 - 17 2597 88

View Full Product Details