Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PUS7
Entrez Gene ID
54517
Gene Name
pseudouridylate synthase 7 (putative)
Gene Aliases
-
Chromosome Location
Chr.7: 105456501 - 105522270 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318163.1 NP_001305092.1 11 - 12 1418 132
NM_001318164.1 NP_001305093.1 11 - 12 1607 132
NM_019042.4 NP_061915.2 11 - 12 1631 132
XM_005250462.3 XP_005250519.1 12 - 13 1657 132
XM_011516336.2 XP_011514638.1 12 - 13 1652 132
XM_017012367.1 XP_016867856.1 12 - 13 1585 132
XM_017012368.1 XP_016867857.1 10 - 11 1498 132
GenBank mRNA AB067484.1 - 11 - 12 1345 132
AK000492.1 - 1 - 2 135 132
AK091283.1 - 11 - 12 1611 132
AK128629.1 - 11 - 12 1593 132
BC005209.2 - 3 - 4 344 132
BC011396.1 - 11 - 12 1587 132

View Full Product Details