The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318163.1 | NP_001305092.1 | 8 - 9 | 1074 | 68 |
NM_001318164.1 | NP_001305093.1 | 8 - 9 | 1263 | 68 | |
NM_019042.4 | NP_061915.2 | 8 - 9 | 1287 | 68 | |
XM_005250462.3 | XP_005250519.1 | 9 - 10 | 1313 | 68 | |
XM_011516336.2 | XP_011514638.1 | 9 - 10 | 1308 | 68 | |
XM_017012367.1 | XP_016867856.1 | 9 - 10 | 1241 | 68 | |
XM_017012368.1 | XP_016867857.1 | 7 - 8 | 1154 | 68 | |
GenBank mRNA | AB067484.1 | - | 8 - 9 | 1001 | 68 |
AK091283.1 | - | 8 - 9 | 1267 | 68 | |
AK128629.1 | - | 8 - 9 | 1249 | 68 | |
BC011396.1 | - | 8 - 9 | 1243 | 68 |