The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306082.1 | NP_001293011.1 | 6 - 7 | 925 | 74 |
NM_018401.2 | NP_060871.1 | 5 - 6 | 815 | 74 | |
XM_017008374.1 | XP_016863863.1 | 4 - 5 | 416 | 74 | |
XM_017008375.1 | XP_016863864.1 | 4 - 5 | 416 | 74 | |
XM_017008376.1 | XP_016863865.1 | 4 - 5 | 416 | 74 | |
GenBank mRNA | AJ250839.1 | - | 5 - 6 | 532 | 74 |
AK292696.1 | - | 5 - 6 | 782 | 74 | |
AK313846.1 | - | 5 - 6 | 817 | 74 | |
AY358353.1 | - | 5 - 6 | 559 | 74 | |
BC038238.1 | - | 5 - 6 | 509 | 74 | |
BC069203.1 | - | 6 - 7 | 846 | 74 |