The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306082.1 | NP_001293011.1 | 10 - 11 | 1364 | 66 |
NM_018401.2 | NP_060871.1 | 9 - 10 | 1254 | 66 | |
XM_005247982.3 | XP_005248039.1 | 4 - 5 | 1761 | 66 | |
XM_005247983.4 | XP_005248040.1 | 4 - 5 | 1019 | 66 | |
XM_011513497.1 | XP_011511799.1 | 8 - 9 | 1358 | 66 | |
XM_011513501.2 | XP_011511803.1 | 5 - 6 | 553 | 66 | |
XM_011513504.1 | XP_011511806.1 | 4 - 5 | 407 | 66 | |
XM_017008377.1 | XP_016863866.1 | 4 - 5 | 1806 | 66 | |
GenBank mRNA | AJ250839.1 | - | 9 - 10 | 971 | 66 |
AK292696.1 | - | 9 - 10 | 1221 | 66 | |
AK313846.1 | - | 9 - 10 | 1256 | 66 | |
AL832436.1 | - | 3 - 4 | 307 | 66 | |
AY358353.1 | - | 9 - 10 | 998 | 66 | |
BC038238.1 | - | 9 - 10 | 948 | 66 | |
BC069203.1 | - | 10 - 11 | 1285 | 66 |