Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CCL4
- Entrez Gene ID
-
6351
- Gene Name
- C-C motif chemokine ligand 4
- Gene Aliases
-
ACT2, AT744.1, G-26, HC21, LAG-1, LAG1, MIP-1-beta, MIP1B, MIP1B1, SCYA2, SCYA4
- Chromosome Location
-
Chr.17: 36103827 - 36105621 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY312578.1 |
- |
1 - 2 |
89 |
96 |
AY766448.1 |
- |
1 - 2 |
157 |
96 |
View Full Product Details