The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017750.3 | NP_060220.3 | 7 - 8 | 1453 | 94 |
GenBank mRNA | AK000303.1 | - | 5 - 6 | 682 | 94 |
AK075261.1 | - | 7 - 8 | 1287 | 94 | |
AK225305.1 | - | 5 - 6 | 682 | 94 | |
AK296522.1 | - | 6 - 7 | 1107 | 94 | |
AL833237.1 | - | 7 - 8 | 1269 | 94 | |
AY358568.1 | - | 7 - 8 | 1275 | 94 | |
BC011418.1 | - | 5 - 6 | 783 | 94 | |
DB001412.1 | - | 2 - 3 | 206 | 94 |