Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DQX1
Entrez Gene ID
165545
Gene Name
DEAQ-box RNA dependent ATPase 1
Gene Aliases
-
Chromosome Location
Chr.2: 74518131 - 74526281 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133637.2 NP_598376.2 4 - 5 979 75
XM_011532644.1 XP_011530946.1 3 - 4 658 75
XM_011532646.2 XP_011530948.1 4 - 5 979 75
XM_017003520.1 XP_016859009.1 2 - 3 432 75
GenBank mRNA AK074337.1 - 4 - 5 886 75
AK092420.1 - 4 - 5 980 75
BC075018.2 - 2 - 3 506 75

View Full Product Details