Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DAP3
Entrez Gene ID
7818
Gene Name
death associated protein 3
Gene Aliases
DAP-3, MRP-S29, MRPS29, bMRP-10
Chromosome Location
Chr.1: 155688104 - 155739010 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199849.1 NP_001186778.1 5 - 6 454 91
NM_001199851.1 NP_001186780.1 3 - 4 238 91
NM_004632.3 NP_004623.1 4 - 5 358 91
NM_033657.2 NP_387506.1 4 - 5 427 91
XM_005245481.2 XP_005245538.1 3 - 4 303 91
XM_011509972.2 XP_011508274.1 4 - 5 357 91
XM_017002289.1 XP_016857778.1 4 - 5 426 91
XM_017002290.1 XP_016857779.1 5 - 6 450 91
XM_017002292.1 XP_016857781.1 4 - 5 323 91
XM_017002293.1 XP_016857782.1 3 - 4 242 91
XM_017002295.1 XP_016857784.1 4 - 5 323 91
XM_017002296.1 XP_016857785.1 3 - 4 303 91
GenBank mRNA AK225869.1 - 5 - 6 454 91
AK293117.1 - 4 - 5 359 91
AK298201.1 - 3 - 4 246 91
BC002358.1 - 3 - 4 386 91
BC107487.1 - 4 - 5 358 91
BC107488.1 - 4 - 5 427 91
BT019494.1 - 3 - 4 270 91
CR749790.1 - 4 - 5 438 91
U18321.1 - 4 - 5 439 91
X83544.1 - 4 - 5 343 91

View Full Product Details