Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VWA5B1
Entrez Gene ID
127731
Gene Name
von Willebrand factor A domain containing 5B1
Gene Aliases
-
Chromosome Location
Chr.1: 20290907 - 20360442 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039500.2 NP_001034589.2 19 - 20 3147 124
XM_006710360.3 XP_006710423.2 16 - 17 2422 124
XM_011540683.2 XP_011538985.1 19 - 20 3109 124
XM_011540684.2 XP_011538986.1 19 - 20 3234 124
XM_011540685.2 XP_011538987.1 19 - 20 3246 124
XM_011540690.2 XP_011538992.1 18 - 19 2932 124
XM_011540691.2 XP_011538993.1 18 - 19 2923 124
XM_011540692.2 XP_011538994.1 19 - 20 3103 124
XM_011540697.2 XP_011538999.1 13 - 14 2480 124
XM_017000305.1 XP_016855794.1 13 - 14 2226 124
XM_017000306.1 XP_016855795.1 14 - 15 2387 124
GenBank mRNA AK057346.1 - 9 - 10 1431 124
AK295797.1 - 19 - 20 3131 124
BC101378.1 - 8 - 9 1227 124
BC101380.1 - 8 - 9 1227 124
BC101381.1 - 8 - 9 1227 124

View Full Product Details