Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VWA5B1
Entrez Gene ID
127731
Gene Name
von Willebrand factor A domain containing 5B1
Gene Aliases
-
Chromosome Location
Chr.1: 20290907 - 20360442 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039500.2 NP_001034589.2 6 - 7 1040 74
XM_006710360.3 XP_006710423.2 3 - 4 315 74
XM_011540683.2 XP_011538985.1 6 - 7 1002 74
XM_011540684.2 XP_011538986.1 6 - 7 1127 74
XM_011540685.2 XP_011538987.1 6 - 7 1139 74
XM_011540688.2 XP_011538990.1 6 - 7 1002 74
XM_011540690.2 XP_011538992.1 6 - 7 1002 74
XM_011540691.2 XP_011538993.1 6 - 7 1002 74
XM_011540692.2 XP_011538994.1 6 - 7 996 74
XM_011540693.1 XP_011538995.1 6 - 7 1002 74
XM_011540695.1 XP_011538997.1 6 - 7 1002 74
XM_011540698.1 XP_011539000.1 6 - 7 1002 74
GenBank mRNA AK125833.1 - 6 - 7 1188 74
AK295797.1 - 6 - 7 1024 74

View Full Product Details