The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039500.2 | NP_001034589.2 | 7 - 8 | 1157 | 94 |
XM_006710360.3 | XP_006710423.2 | 4 - 5 | 432 | 94 | |
XM_011540683.2 | XP_011538985.1 | 7 - 8 | 1119 | 94 | |
XM_011540684.2 | XP_011538986.1 | 7 - 8 | 1244 | 94 | |
XM_011540685.2 | XP_011538987.1 | 7 - 8 | 1256 | 94 | |
XM_011540688.2 | XP_011538990.1 | 7 - 8 | 1119 | 94 | |
XM_011540691.2 | XP_011538993.1 | 7 - 8 | 1119 | 94 | |
XM_011540692.2 | XP_011538994.1 | 7 - 8 | 1113 | 94 | |
XM_011540693.1 | XP_011538995.1 | 7 - 8 | 1119 | 94 | |
XM_011540695.1 | XP_011538997.1 | 7 - 8 | 1119 | 94 | |
XM_011540698.1 | XP_011539000.1 | 7 - 8 | 1119 | 94 | |
GenBank mRNA | AK295797.1 | - | 7 - 8 | 1141 | 94 |