The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163484.1 | NP_001156956.1 | 7 - 8 | 997 | 84 |
NM_025230.4 | NP_079506.3 | 7 - 8 | 1447 | 84 | |
NM_181357.2 | NP_852002.1 | 7 - 8 | 799 | 84 | |
NR_028099.1 | 6 - 7 | 1359 | 84 | ||
NR_028100.1 | 6 - 7 | 974 | 84 | ||
GenBank mRNA | AB209073.1 | - | 6 - 7 | 1345 | 84 |
AF130070.1 | - | 6 - 7 | 707 | 84 | |
AF267858.1 | - | 7 - 8 | 1110 | 84 | |
AK057636.1 | - | 7 - 8 | 758 | 84 | |
AK074296.1 | - | 7 - 8 | 1412 | 84 | |
AK292797.1 | - | 7 - 8 | 879 | 84 | |
AK296742.1 | - | 6 - 7 | 931 | 84 | |
AK307849.1 | - | 4 - 5 | 579 | 84 | |
BC008858.2 | - | 2 - 3 | 225 | 84 | |
BC067132.1 | - | 7 - 8 | 870 | 84 | |
BX161405.1 | - | 6 - 7 | 694 | 84 | |
BX247970.1 | - | 5 - 6 | 873 | 84 | |
BX648224.1 | - | 7 - 8 | 929 | 84 |