The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163484.1 | NP_001156956.1 | 3 - 4 | 563 | 70 |
NM_025230.4 | NP_079506.3 | 3 - 4 | 1013 | 70 | |
NM_181357.2 | NP_852002.1 | 3 - 4 | 365 | 70 | |
NR_028099.1 | 3 - 4 | 990 | 70 | ||
NR_028100.1 | 2 - 3 | 540 | 70 | ||
GenBank mRNA | AB209073.1 | - | 3 - 4 | 976 | 70 |
AF130070.1 | - | 2 - 3 | 273 | 70 | |
AF267858.1 | - | 3 - 4 | 676 | 70 | |
AK057636.1 | - | 3 - 4 | 324 | 70 | |
AK074296.1 | - | 3 - 4 | 978 | 70 | |
AK292797.1 | - | 3 - 4 | 445 | 70 | |
AK296742.1 | - | 2 - 3 | 497 | 70 | |
BC067132.1 | - | 3 - 4 | 436 | 70 | |
BX161405.1 | - | 2 - 3 | 260 | 70 | |
BX247970.1 | - | 2 - 3 | 504 | 70 | |
BX648224.1 | - | 3 - 4 | 495 | 70 | |
DB212727.1 | - | 3 - 4 | 446 | 70 |