Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DCAF11
Entrez Gene ID
80344
Gene Name
DDB1 and CUL4 associated factor 11
Gene Aliases
GL014, PRO2389, WDR23
Chromosome Location
Chr.14: 24114697 - 24125242 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA CD644652.1 - 1 - 2 147 69

View Full Product Details