Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INPP5B
Entrez Gene ID
3633
Gene Name
inositol polyphosphate-5-phosphatase B
Gene Aliases
5PTase
Chromosome Location
Chr.1: 37860697 - 37947094 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297434.1 NP_001284363.1 8 - 9 1020 143
NM_005540.2 NP_005531.2 14 - 15 1527 143
XM_006710621.3 XP_006710684.2 10 - 11 1290 143
XM_011541392.2 XP_011539694.1 8 - 9 966 143
XM_011541393.2 XP_011539695.1 8 - 9 955 143
XM_017001206.1 XP_016856695.1 14 - 15 1564 143
XM_017001207.1 XP_016856696.1 11 - 12 1267 143
XM_017001208.1 XP_016856697.1 11 - 12 1396 143
XM_017001209.1 XP_016856698.1 8 - 9 946 143
GenBank mRNA AK022846.1 - 8 - 9 978 143
AK310824.1 - 14 - 15 1517 143
BC042529.2 - 7 - 8 902 143
BC058932.1 - 14 - 15 1491 143
M74161.1 - 14 - 15 1521 143

View Full Product Details