The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006343.2 | NP_006334.2 | 8 - 9 | 1417 | 55 |
XM_005263565.4 | XP_005263622.1 | 8 - 9 | 1552 | 55 | |
XM_005263568.4 | XP_005263625.1 | 8 - 9 | 1552 | 55 | |
XM_011510490.2 | XP_011508792.1 | 8 - 9 | 1452 | 55 | |
XM_017003164.1 | XP_016858653.1 | 8 - 9 | 1422 | 55 | |
XM_017003165.1 | XP_016858654.1 | 9 - 10 | 1521 | 55 | |
GenBank mRNA | AK316584.1 | - | 8 - 9 | 1424 | 55 |
BC114917.1 | - | 8 - 9 | 1131 | 55 | |
BC114918.1 | - | 8 - 9 | 1131 | 55 | |
U08023.1 | - | 8 - 9 | 1432 | 55 |