The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322438.1 | NP_001309367.1 | 4 - 5 | 576 | 149 |
NM_001322439.1 | NP_001309368.1 | 4 - 5 | 639 | 149 | |
NM_001322440.1 | NP_001309369.1 | 4 - 5 | 601 | 149 | |
NM_001322441.1 | NP_001309370.1 | 4 - 5 | 704 | 149 | |
NM_001322451.1 | NP_001309380.1 | 3 - 4 | 462 | 149 | |
NM_001322452.1 | NP_001309381.1 | 3 - 4 | 481 | 149 | |
NM_001322453.1 | NP_001309382.1 | 3 - 4 | 500 | 149 | |
NM_021644.3 | NP_067676.2 | 4 - 5 | 620 | 149 | |
GenBank mRNA | AF132361.1 | - | 4 - 5 | 509 | 149 |
AK291547.1 | - | 4 - 5 | 572 | 149 | |
BC039824.1 | - | 4 - 5 | 556 | 149 |