The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282501.1 | NP_001269430.1 | 3 - 4 | 319 | 61 |
NM_175743.2 | NP_786885.1 | 4 - 5 | 398 | 61 | |
GenBank mRNA | AK290443.1 | - | 3 - 4 | 319 | 61 |
BC063681.1 | - | 2 - 3 | 185 | 61 | |
BG575302.1 | - | 4 - 5 | 343 | 61 | |
BX282627.1 | - | 4 - 5 | 358 | 61 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321400.1 | NP_001308329.1 | 4 - 5 | 398 | 61 |
NM_001321402.1 | NP_001308331.1 | 3 - 4 | 319 | 61 |