The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145001.2 | NP_001138473.1 | 4 - 5 | 595 | 57 |
NM_001166167.1 | NP_001159639.1 | 3 - 4 | 377 | 57 | |
NM_001166168.1 | NP_001159640.1 | 3 - 4 | 372 | 57 | |
NM_001166169.1 | NP_001159641.1 | 3 - 4 | 381 | 57 | |
NM_001166170.1 | NP_001159642.1 | 3 - 4 | 379 | 57 | |
NM_001166171.1 | NP_001159643.1 | 4 - 5 | 544 | 57 | |
NM_014397.5 | NP_055212.2 | 3 - 4 | 430 | 57 | |
XM_005251664.1 | XP_005251721.1 | 3 - 4 | 436 | 57 | |
XM_006716936.2 | XP_006716999.1 | 3 - 4 | 441 | 57 | |
XM_017014217.1 | XP_016869706.1 | 4 - 5 | 604 | 57 | |
GenBank mRNA | AB026289.1 | - | 3 - 4 | 361 | 57 |
AF087909.1 | - | 3 - 4 | 366 | 57 | |
AK294614.1 | - | 3 - 4 | 381 | 57 | |
AK313071.1 | - | 3 - 4 | 331 | 57 | |
BC000101.2 | - | 3 - 4 | 376 | 57 | |
BC004174.2 | - | 4 - 5 | 548 | 57 | |
BC004209.2 | - | 4 - 5 | 548 | 57 | |
BC012761.2 | - | 3 - 4 | 377 | 57 | |
BM083872.1 | - | 3 - 4 | 377 | 57 | |
BP265230.1 | - | 3 - 4 | 381 | 57 | |
BP279761.1 | - | 3 - 4 | 372 | 57 | |
CF994196.1 | - | 3 - 4 | 457 | 57 | |
CR457091.1 | - | 2 - 3 | 211 | 57 | |
CR542222.1 | - | 2 - 3 | 211 | 57 |