Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTCD2
Entrez Gene ID
79810
Gene Name
pentatricopeptide repeat domain 2
Gene Aliases
-
Chromosome Location
Chr.5: 72320367 - 72368395 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284404.1 NP_001271333.1 2 - 3 232 84
NM_001284405.1 NP_001271334.1 2 - 3 232 84
NM_024754.4 NP_079030.3 2 - 3 232 84
XM_005248601.2 XP_005248658.1 2 - 3 226 84
XM_017009866.1 XP_016865355.1 2 - 3 211 84
GenBank mRNA AK056761.1 - 2 - 3 896 84
AK303639.1 - 2 - 3 201 84
AK303776.1 - 2 - 3 231 84
BC018720.1 - 2 - 3 208 84
DA082506.1 - 2 - 3 226 84
DC393817.1 - 2 - 3 232 84

View Full Product Details