The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284403.1 | NP_001271332.1 | 3 - 4 | 331 | 132 |
NM_001284404.1 | NP_001271333.1 | 4 - 5 | 461 | 132 | |
NM_001284405.1 | NP_001271334.1 | 5 - 6 | 579 | 132 | |
NM_024754.4 | NP_079030.3 | 6 - 7 | 658 | 132 | |
XM_005248601.2 | XP_005248658.1 | 6 - 7 | 652 | 132 | |
XM_005248603.2 | XP_005248660.1 | 5 - 6 | 514 | 132 | |
XM_017009866.1 | XP_016865355.1 | 5 - 6 | 519 | 132 | |
XM_017009867.1 | XP_016865356.1 | 4 - 5 | 431 | 132 | |
XM_017009868.1 | XP_016865357.1 | 5 - 6 | 514 | 132 | |
GenBank mRNA | AK022660.1 | - | 4 - 5 | 420 | 132 |
AK056761.1 | - | 6 - 7 | 1322 | 132 | |
AK298750.1 | - | 3 - 4 | 322 | 132 | |
AK303639.1 | - | 4 - 5 | 430 | 132 | |
AK303776.1 | - | 5 - 6 | 578 | 132 | |
BC018720.1 | - | 6 - 7 | 634 | 132 | |
CR457290.1 | - | 2 - 3 | 186 | 132 |