The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204526.1 | NP_001191455.1 | 4 - 5 | 411 | 65 |
NM_001204527.1 | NP_001191456.1 | 4 - 5 | 434 | 65 | |
NM_006280.2 | NP_006271.1 | 3 - 4 | 384 | 65 | |
NR_037927.1 | 5 - 6 | 605 | 65 | ||
XM_011531186.1 | XP_011529488.1 | 4 - 5 | 293 | 65 | |
XM_011531187.1 | XP_011529489.1 | 4 - 5 | 293 | 65 | |
XM_017029756.1 | XP_016885245.1 | 4 - 5 | 1182 | 65 | |
XM_017029757.1 | XP_016885246.1 | 4 - 5 | 917 | 65 | |
GenBank mRNA | AK057117.1 | - | 2 - 3 | 1967 | 65 |
AK094395.1 | - | 3 - 4 | 2087 | 65 | |
AK290493.1 | - | 3 - 4 | 290 | 65 | |
BC003371.1 | - | 3 - 4 | 264 | 65 | |
BE791319.1 | - | 3 - 4 | 306 | 65 | |
BG165852.1 | - | 4 - 5 | 418 | 65 | |
BP202705.1 | - | 4 - 5 | 373 | 65 | |
BT007192.1 | - | 3 - 4 | 260 | 65 | |
CD672615.1 | - | 4 - 5 | 476 | 65 | |
DB238237.1 | - | 3 - 4 | 383 | 65 | |
X90583.1 | - | 3 - 4 | 290 | 65 | |
Z69043.1 | - | 3 - 4 | 309 | 65 |