Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRSF7
Entrez Gene ID
6432
Gene Name
serine and arginine rich splicing factor 7
Gene Aliases
9G8, AAG3, SFRS7
Chromosome Location
Chr.2: 38743599 - 38751494 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK091425.1 - 3 - 4 444 63

View Full Product Details