The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135191.1 | NP_001128663.1 | 26 - 27 | 3157 | 100 |
NM_003887.2 | NP_003878.1 | 27 - 28 | 3292 | 100 | |
XM_006711898.1 | XP_006711961.1 | 28 - 29 | 3054 | 100 | |
XM_006711901.3 | XP_006711964.1 | 28 - 29 | 2904 | 100 | |
XM_006711902.1 | XP_006711965.1 | 27 - 28 | 2919 | 100 | |
XM_011510403.2 | XP_011508705.1 | 28 - 29 | 3075 | 100 | |
XM_011510404.2 | XP_011508706.1 | 27 - 28 | 3065 | 100 | |
XM_011510405.2 | XP_011508707.1 | 27 - 28 | 3001 | 100 | |
XM_011510406.2 | XP_011508708.1 | 29 - 30 | 3008 | 100 | |
XM_011510407.2 | XP_011508709.1 | 27 - 28 | 2940 | 100 | |
XM_011510409.1 | XP_011508711.1 | 28 - 29 | 2946 | 100 | |
GenBank mRNA | AB007860.1 | - | 27 - 28 | 3292 | 100 |
AK091121.1 | - | 5 - 6 | 703 | 100 | |
AK124900.1 | - | 3 - 4 | 1450 | 100 | |
BC063308.1 | - | 26 - 27 | 3110 | 100 |