Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FMO1
Entrez Gene ID
2326
Gene Name
flavin containing monooxygenase 1
Gene Aliases
-
Chromosome Location
Chr.1: 171248471 - 171285978 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005245038.3 XP_005245095.1 7 - 8 1348 121

View Full Product Details