The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018441.5 | NP_060911.2 | 1 - 2 | 196 | 68 |
GenBank mRNA | AF212234.1 | - | 1 - 2 | 181 | 68 |
AF232009.1 | - | 1 - 2 | 186 | 68 | |
AJ250303.1 | - | 1 - 2 | 182 | 68 | |
AK296208.1 | - | 1 - 2 | 170 | 68 | |
AK315795.1 | - | 1 - 2 | 155 | 68 | |
BC002529.2 | - | 1 - 2 | 160 | 68 | |
BU151224.1 | - | 1 - 2 | 196 | 68 | |
BX471043.1 | - | 1 - 2 | 190 | 68 |