The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003872.2 | NP_003863.2 | 6 - 7 | 1778 | 96 |
NM_018534.3 | NP_061004.3 | 6 - 7 | 1778 | 96 | |
NM_201264.1 | NP_957716.1 | 6 - 7 | 1778 | 96 | |
NM_201266.1 | NP_957718.1 | 6 - 7 | 1778 | 96 | |
NM_201267.1 | NP_957719.1 | 6 - 7 | 1778 | 96 | |
NM_201279.1 | NP_958436.1 | 6 - 7 | 1778 | 96 | |
XM_005246933.3 | XP_005246990.2 | 7 - 8 | 1643 | 96 | |
XM_005246934.3 | XP_005246991.2 | 7 - 8 | 1316 | 96 | |
XM_017005185.1 | XP_016860674.1 | 7 - 8 | 1642 | 96 | |
XM_017005186.1 | XP_016860675.1 | 7 - 8 | 1643 | 96 | |
XM_017005187.1 | XP_016860676.1 | 6 - 7 | 1828 | 96 | |
XM_017005188.1 | XP_016860677.1 | 6 - 7 | 1827 | 96 | |
GenBank mRNA | AF016098.1 | - | 6 - 7 | 987 | 96 |
AF022859.1 | - | 6 - 7 | 987 | 96 | |
AF022860.1 | - | 6 - 7 | 987 | 96 | |
AF280544.1 | - | 6 - 7 | 987 | 96 | |
AF280545.1 | - | 6 - 7 | 987 | 96 | |
AF280546.1 | - | 6 - 7 | 987 | 96 | |
AK290934.1 | - | 7 - 8 | 1458 | 96 | |
BC101525.1 | - | 6 - 7 | 1018 | 96 | |
BC104770.1 | - | 6 - 7 | 1018 | 96 | |
BC117413.1 | - | 6 - 7 | 1018 | 96 | |
BC143237.1 | - | 6 - 7 | 1018 | 96 | |
BC143238.1 | - | 6 - 7 | 1018 | 96 | |
BC143608.1 | - | 6 - 7 | 1018 | 96 | |
BX537423.1 | - | 6 - 7 | 1777 | 96 | |
KJ534899.1 | - | 6 - 7 | 987 | 96 | |
KJ534900.1 | - | 6 - 7 | 987 | 96 |