The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016355.3 | NP_057439.2 | 8 - 9 | 922 | 96 |
GenBank mRNA | AF190165.1 | - | 7 - 8 | 1435 | 96 |
AK054574.1 | - | 8 - 9 | 907 | 96 | |
AK222940.1 | - | 8 - 9 | 881 | 96 | |
AK225144.1 | - | 8 - 9 | 892 | 96 | |
AK310501.1 | - | 8 - 9 | 952 | 96 | |
AL136666.1 | - | 8 - 9 | 926 | 96 | |
BC009379.2 | - | 8 - 9 | 885 | 96 | |
BC068009.1 | - | 8 - 9 | 904 | 96 |