Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KATNAL1
Entrez Gene ID
84056
Gene Name
katanin catalytic subunit A1 like 1
Gene Aliases
-
Chromosome Location
Chr.13: 30202626 - 30307500 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032116.4 NP_115492.1 1 - 2 164 105
XM_017020791.1 XP_016876280.1 1 - 2 177 105
GenBank mRNA AK291439.1 - 1 - 2 97 105
AK315724.1 - 1 - 2 101 105
BC000612.2 - 1 - 2 124 105
DR006891.1 - 1 - 2 199 105

View Full Product Details