The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014380.2 | NP_001014402.1 | 8 - 9 | 1142 | 103 |
NM_032116.4 | NP_115492.1 | 8 - 9 | 1184 | 103 | |
XM_005266574.2 | XP_005266631.1 | 8 - 9 | 1326 | 103 | |
XM_017020791.1 | XP_016876280.1 | 7 - 8 | 1028 | 103 | |
XM_017020792.1 | XP_016876281.1 | 7 - 8 | 1049 | 103 | |
XM_017020793.1 | XP_016876282.1 | 5 - 6 | 1358 | 103 | |
GenBank mRNA | AK097423.1 | - | 8 - 9 | 1114 | 103 |
AK291439.1 | - | 8 - 9 | 1117 | 103 | |
AK315724.1 | - | 8 - 9 | 1121 | 103 | |
BC000612.2 | - | 8 - 9 | 1144 | 103 |