Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRM1
Entrez Gene ID
79922
Gene Name
mitochondrial rRNA methyltransferase 1
Gene Aliases
-
Chromosome Location
Chr.17: 36601581 - 36634698 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK026231.1 - 1 - 2 216 74
AK225449.1 - 1 - 2 216 74

View Full Product Details