The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001314039.1 | NP_001300968.1 | 5 - 6 | 772 | 83 |
NM_001539.3 | NP_001530.1 | 6 - 7 | 950 | 83 | |
GenBank mRNA | AJ277242.1 | - | 4 - 5 | 558 | 83 |
AK095541.1 | - | 3 - 4 | 83 | ||
AK289623.1 | - | 6 - 7 | 878 | 83 | |
AK298725.1 | - | 5 - 6 | 700 | 83 | |
AY186741.1 | - | 6 - 7 | 876 | 83 | |
BC008182.1 | - | 6 - 7 | 863 | 83 | |
BT007292.1 | - | 5 - 6 | 759 | 83 | |
D13388.1 | - | 6 - 7 | 876 | 83 | |
L08069.1 | - | 6 - 7 | 841 | 83 |