Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CDYL
- Entrez Gene ID
-
9425
- Gene Name
- chromodomain protein, Y-like
- Gene Aliases
-
CDYL1
- Chromosome Location
-
Chr.6: 4706159 - 4955544 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_026590.1 |
|
1 - 2 |
|
120 |
GenBank mRNA |
AF081258.1 |
- |
1 - 2 |
88 |
120 |
AF081259.1 |
- |
1 - 2 |
88 |
120 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC497048 |
KU-MEL-3 |
|
-94.2kb
|
Sense |
Chr. - 4610565 - 4611919 on Build GRCh38 |
Downstream |
ribonuclease P/MRP subunit p40 |
RPP40 |
RNASEP1, bA428J1.3 |
276.5kb
|
Anti-sense |
Chr. - 4992405 - 5004063 on Build GRCh38 |
View Full Product Details