The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243746.1 | NP_001230675.1 | 7 - 8 | 1448 | 62 |
NM_017565.3 | NP_060035.2 | 6 - 7 | 1222 | 62 | |
NR_027751.1 | 6 - 7 | 624 | 62 | ||
XM_006721959.2 | XP_006722022.1 | 6 - 7 | 594 | 62 | |
XM_011524918.2 | XP_011523220.1 | 6 - 7 | 1062 | 62 | |
XM_017024781.1 | XP_016880270.1 | 6 - 7 | 1062 | 62 | |
GenBank mRNA | AB545606.1 | - | 6 - 7 | 1222 | 62 |
AK056789.1 | - | 7 - 8 | 1062 | 62 | |
AK298071.1 | - | 7 - 8 | 1090 | 62 | |
AL133105.1 | - | 4 - 5 | 331 | 62 | |
AY358197.1 | - | 7 - 8 | 1222 | 62 | |
BC036222.1 | - | 6 - 7 | 588 | 62 | |
BC136686.1 | - | 6 - 7 | 1111 | 62 | |
BC136689.1 | - | 7 - 8 | 1111 | 62 |