The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031684.2 | NP_001026854.1 | 7 - 8 | 902 | 75 |
XM_005264484.2 | XP_005264541.1 | 7 - 8 | 760 | 75 | |
XM_011533032.2 | XP_011531334.1 | 5 - 6 | 901 | 75 | |
GenBank mRNA | AB445102.1 | - | 7 - 8 | 664 | 75 |
AK091425.1 | - | 8 - 9 | 1166 | 75 | |
AK225141.1 | - | 6 - 7 | 679 | 75 | |
AY166860.1 | - | 7 - 8 | 790 | 75 | |
AY513287.1 | - | 7 - 8 | 745 | 75 | |
BC000997.2 | - | 7 - 8 | 749 | 75 | |
BC017369.2 | - | 7 - 8 | 760 | 75 | |
BC017908.1 | - | 7 - 8 | 767 | 75 | |
BC022328.1 | - | 7 - 8 | 767 | 75 | |
BT006745.1 | - | 7 - 8 | 664 | 75 | |
L22253.1 | - | 7 - 8 | 769 | 75 |