The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142643.2 | NP_001136115.1 | 11 - 12 | 1384 | 105 |
NM_020753.4 | NP_065804.2 | 12 - 13 | 1901 | 105 | |
GenBank mRNA | AB032965.1 | - | 8 - 9 | 1080 | 105 |
AF451976.1 | - | 12 - 13 | 1754 | 105 | |
AK300354.1 | - | 11 - 12 | 1331 | 105 | |
AK315936.1 | - | 11 - 12 | 1385 | 105 | |
BC066643.1 | - | 12 - 13 | 1864 | 105 |