Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CASKIN2
Entrez Gene ID
57513
Gene Name
CASK interacting protein 2
Gene Aliases
ANKS5B
Chromosome Location
Chr.17: 75500260 - 75515583 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142643.2 NP_001136115.1 7 - 8 793 85
NM_020753.4 NP_065804.2 8 - 9 1310 85
GenBank mRNA AB032965.1 - 4 - 5 489 85
AF451976.1 - 8 - 9 1163 85
AK300354.1 - 7 - 8 740 85
AK315936.1 - 7 - 8 794 85
BC066643.1 - 8 - 9 1273 85

View Full Product Details