The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032711.3 | NP_116100.2 | 1 - 2 | 220 | 128 |
GenBank mRNA | BC012327.1 | - | 1 - 2 | 184 | 128 |
U84249.1 | - | 1 - 2 | 106 | 128 |