The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042559.2 | NP_001036024.3 | 17 - 18 | 2421 | 128 |
NM_001172705.1 | NP_001166176.1 | 18 - 19 | 2652 | 128 | |
NM_001418.3 | NP_001409.3 | 18 - 19 | 2535 | 128 | |
GenBank mRNA | AB209267.1 | - | 18 - 19 | 2413 | 128 |
AK223548.1 | - | 18 - 19 | 2456 | 128 | |
AK302886.1 | - | 19 - 20 | 2363 | 128 | |
AY513274.1 | - | 18 - 19 | 2419 | 128 | |
BC014930.2 | - | 2 - 3 | 455 | 128 | |
BC018746.1 | - | 18 - 19 | 2413 | 128 | |
BC018975.2 | - | 18 - 19 | 2402 | 128 | |
BC039851.1 | - | 18 - 19 | 2418 | 128 | |
BC043149.1 | - | 18 - 19 | 2431 | 128 | |
BC111415.1 | - | 17 - 18 | 2208 | 128 | |
BC111548.1 | - | 16 - 17 | 2094 | 128 | |
U73824.1 | - | 18 - 19 | 2424 | 128 | |
U76111.1 | - | 18 - 19 | 2425 | 128 | |
X89713.1 | - | 17 - 18 | 2157 | 128 |