The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042559.2 | NP_001036024.3 | 19 - 20 | 2841 | 157 |
NM_001172705.1 | NP_001166176.1 | 20 - 21 | 3072 | 157 | |
NM_001418.3 | NP_001409.3 | 20 - 21 | 2955 | 157 | |
GenBank mRNA | AB063323.1 | - | 3 - 4 | 506 | 157 |
AB209267.1 | - | 20 - 21 | 2833 | 157 | |
AK223548.1 | - | 20 - 21 | 2876 | 157 | |
AK302886.1 | - | 21 - 22 | 2783 | 157 | |
AY513274.1 | - | 20 - 21 | 2839 | 157 | |
BC014930.2 | - | 4 - 5 | 875 | 157 | |
BC018746.1 | - | 20 - 21 | 2833 | 157 | |
BC018975.2 | - | 20 - 21 | 2822 | 157 | |
BC039851.1 | - | 20 - 21 | 2838 | 157 | |
BC043149.1 | - | 20 - 21 | 2851 | 157 | |
BC065276.1 | - | 20 - 21 | 3049 | 157 | |
BC111415.1 | - | 19 - 20 | 2628 | 157 | |
BC111548.1 | - | 18 - 19 | 2514 | 157 | |
BX647799.1 | - | 13 - 14 | 5897 | 157 | |
U73824.1 | - | 20 - 21 | 2844 | 157 | |
U76111.1 | - | 20 - 21 | 2845 | 157 | |
X89713.1 | - | 19 - 20 | 2577 | 157 |