Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EIF4G2
Entrez Gene ID
1982
Gene Name
eukaryotic translation initiation factor 4 gamma 2
Gene Aliases
AAG1, DAP5, NAT1, P97
Chromosome Location
Chr.11: 10797046 - 10809035 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042559.2 NP_001036024.3 1 - 2 333 109
NM_001418.3 NP_001409.3 1 - 2 333 109
GenBank mRNA AB209267.1 - 1 - 2 211 109
AK223548.1 - 1 - 2 254 109
AY513274.1 - 1 - 2 217 109
BC018746.1 - 1 - 2 211 109
BC018975.2 - 1 - 2 200 109
BC039851.1 - 1 - 2 216 109
BC043149.1 - 1 - 2 229 109
DB072494.1 - 1 - 2 333 109
U73824.1 - 1 - 2 222 109
U76111.1 - 1 - 2 110

View Full Product Details