The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012192.3 | NP_036324.1 | 1 - 2 | 111 | 61 |
GenBank mRNA | AF150105.1 | - | 1 - 2 | 100 | 61 |
AF152355.1 | - | 1 - 2 | 41 | 61 | |
AF183415.1 | - | 1 - 2 | 99 | 61 | |
AK002130.1 | - | 1 - 2 | 72 | 61 | |
AK311763.1 | - | 1 - 2 | 61 | ||
BC011014.1 | - | 1 - 2 | 53 | 61 | |
DA290694.1 | - | 1 - 2 | 111 | 61 |